Learn More
Greiner Bio-One TC Treated Terasaki Plate
Suitable for all applications for serological determination of HLA antigens. Greiner Bio-One TC Treated Terasaki Plates are supplied with fitting lid.
Brand: Greiner Bio-One 653190
This item is not returnable.
View return policy
Description
- Manufactured from crystal clear polystyrene
- TC surface treatment for improved cell adhesion