Learn More
Greiner Bio-One TC Treated Terasaki Plate
GREENER_CHOICE
Suitable for all applications for serological determination of HLA antigens. Greiner Bio-One TC Treated Terasaki Plates are supplied with fitting lid.
Brand:  Greiner Bio-One 653190
Product Code. 11905325
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products »
Description
Description
  • Manufactured from crystal clear polystyrene
  • TC surface treatment for improved cell adhesion