missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ TC-S 7006

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
TC-S 7006 is a potent and selective Tpl2 (Cot; MAP3K8) inhibitor (IC50 = 50 nM). Selective for Tpl2 over MEK, p38, Src, MK2, PKC and EGFR. Inhibits LPS-induced TNF-α secretion from monocytes and attenuates acute myeloid leukemia (AML) cell proliferation in vitro. Also reduces cytolytic activity of human CD8+ cytotoxic T lymphocytes. Cell permeable.
Specifications
Specifications
| Quantity | 50 mg |
| Product Type | TC-S 7006 |
| Purity | >99% |
| Molecular Weight (g/mol) | 404.83 |
| Inhibitors | TC-S 7006 |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction