missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ TC-S 7006
GREENER_CHOICE

Product Code. 18787668
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18787668

Brand: Tocris Bioscience™ 5240/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent and selective Tpl2 (Cot; MAP3K8) inhibitor

TC-S 7006 is a potent and selective Tpl2 (Cot; MAP3K8) inhibitor (IC50 = 50 nM). Selective for Tpl2 over MEK, p38, Src, MK2, PKC and EGFR. Inhibits LPS-induced TNF-α secretion from monocytes and attenuates acute myeloid leukemia (AML) cell proliferation in vitro. Also reduces cytolytic activity of human CD8+ cytotoxic T lymphocytes. Cell permeable.
TRUSTED_SUSTAINABILITY

Specifications

Quantity 10 mg
Product Type TC-S 7006
Purity >99%
Molecular Weight (g/mol) 404.83
Inhibitors TC-S 7006

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.