Learn More
Cytiva TC-Gasket
Shop All Cytiva Products
Click to view available options
Diameter (Metric) Inner:
4 mm
6.5 mm
Unit Size:
Each
Specifications
Specifications
| Certifications/Compliance | USP Class VI |
| Connection Type | 25 mm |
| Material | Perfluororubber |
| Type | TC-Gasket |
| Quantity | 4 Pack |
| Diameter (Metric) Inner | 6.5 mm |