missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Tazemetostat

Product Code. 30226369
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 30226369

Brand: Tocris Bioscience™ 8093/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent and selective EZH2 inhibitor

Tazemetostat is a potent and selective EZH2 inhibitor (Ki = 2.5 nM; IC50 = 11 nM). Reduces H3K27me3 levels in PDX derived cells and inhibits cellular H3K27 methylation. Tazemetostat suppresses EZH2 and MYC expression and induces neuronal differentiation genes; also increases neurite extension in MB-39-nu cells. Antiproliferative in SMARCAB1-deleted MRT cell lines (IC50 = 32 - 1000 nM); inhibits methylation in both mutant and wild type cells (IC50 = 1.4-4.9 nM). Dose-dependently suppresses growth of both MC38 cells and tumors in vivo.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-[(1,2-Dihydro-4,6-dimethyl-2-oxo-3-pyridinyl)methyl]-5-[ethyl(tetrahydro-2H-pyran-4-yl)amino]-4-methyl-4'-(4-morpholinylmethyl)[1,1'-biphenyl]-3-carboxamide
CAS 1403254-99-8
Quantity 50 mg
Target EZH2 Inhibitor
Molecular Formula C34H44N4O4
Purity 0.97
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.