Learn More
BD Taxo™ Differentiation Discs: Antibiotic - Hippurate
Description
- Organisms react with chemical agents impregnated on discs resulting in positive or negative reactions
- Combination of positive and negative reaction aids in the identification of the organism
Specifications
Specifications
| Antimicrobial Agents | Hippurate |
| Content And Storage | -20°C to +8°C |
| Packaging Type | Disc |
| Quantity | 1 Pack |
For in vitro Diagnostic use. Follow established laboratory procedures in handling and disposing of infectious materials.