Learn More
Novus Biologicals™ Tat-Beclin 1 L11 Autophagy Inducing Peptide
Product Code. 18617916 Shop All Bio Techne Products
Click to view available options
Quantity:
0.1 mg
1 mg
5 mg
Unit Size:
0.1mg
1mg
5mg
Product Code. 18617916
Brand: Novus Biologicals™ NBP2498865mg
Inducing Peptide
TRUSTED_SUSTAINABILITY
Specifications
Species Non-species Specific
Content And Storage Store at -20°C in powder form. Store at -80°C once reconstituted.
Gene Alias Autophagy Inducing peptide, Tat-Beclin, Tat-Beclin 1, Tat-Beclin 1 peptide, Tat-Beclin peptide, Tat-L11
Molecular Weight (g/mol) MolecularWeight-theroretical: 3.08 kDa
Quantity 5 mg
Research Category Autophagy
Storage Buffer This product is supplied lyophilized. Purity is ≥ to 97% (HPLC)