missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ TaqMan™ OpenArray™ Loading Plate, QuantStudio™ 12K Flex
Designed to provide new and experienced users a single OpenArray™ plate to practice the workflow prior to starting custom projects.
Brand: Applied Biosystems™ 4471227
This item is not returnable.
View return policy
Description
The OpenArray™ loading plate does not contain assays and is used for practicing loading techniques.
Specifications
| 1 plate., store at -20°C | |
| Low | |
| Standard | |
| OpenArray™, TaqMan™ | |
| Real Time PCR (qPCR) |
| Primer-Probe | |
| qPCR | |
| QuantStudio™ 12k Flex | |
| Loading Plate |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction