Learn More
Applied Biosystems™ TaqMan™ Array Human MicroRNA A Cards v2.0
Product Code. 11361453 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
4 pack
Unit Size:
Pack of 4
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11361453 4 pack Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11361453 Supplier Applied Biosystems™ Supplier No. 4398965
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
TaqMan™ Array Human MicroRNA A Cards v2.0
The TaqMan™ Array Human MicroRNA A Card v2.0 contains 384 TaqMan™ MicroRNA Assays enabling accurate quantitation of 377 human microRNAs in as little as 5 hours. Three TaqMan MicroRNA Assay endogenous controls are included to aid in data normalization and one TaqMan™ MicroRNA Assay not related to human is included as a negative control. Details on the included assays is available in the Target List file. Purchase of the Megaplex™ RT Primers, Human Pool A v2.1 is required to run this array. An additional preamplification step is enabled by Megaplex™ PreAmp Primers, Human Pool A v2.1 for situations where sensitivity is of the utmost importance and⁄or sample is limiting.
TRUSTED_SUSTAINABILITY
Specifications
Type Array Human MicroRNA A Card
Format 384-well Microfluidic Card
Species Human
Content And Storage 4°C
Form Dried Down
For Use With (Equipment) AB 7900 HT 384-Well System
Internal Probe Modification MGB (Minor Groove Binder) (3'), Non-Fluorescent Quencher (3'), FAM (5')
Label or Dye FAM
Product Line Megaplex, TaqMan
Quantity 4 pack
Reaction Speed Standard
Research Category Gene Regulation, Epigenetics