Learn More
MP Biomedicals™ Taq-&GO Ready-to-Use PCR Mix
A ready-to-use, optimized 5X reaction mix for PCR
110.24€ - 824.68€
Specifications
Content And Storage | Store at -20°C. |
---|---|
For Use With (Application) | Standard PCR |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11427010
|
MP Biomedicals™
11EPTAG100 |
1mL |
110.24€
1mL |
Estimated Shipment: 04-06-2024
Log in to see stock. |
|||||
11437010
|
MP Biomedicals™
11EPTAG110 |
10 x 1mL |
824.68€
10mL |
Estimated Shipment: 07-06-2024
Log in to see stock. |
|||||
Description
Simple-to-use: add 10μL of Taq-&GO to a 40μL volume containing desired template and primers and reaction mix is ready.
Unit Definition: One unit incorporates 10nmol of total dNTPs into acid-precipitable DNA in 30 min. at 74°C.
Specifications
Store at -20°C. | |
Standard PCR |