Learn More
DWK Life Sciences Tamper Evident Screw Cap, Narrow Mouth, Blue PP, for Soda-Lime Glass Screw-Top Bottles
Shop All DWK Life Sciences Products
Click to view available options
Thread Size (Metric):
GL 32
GL 45
Unit Size:
Pack of 10
Description
Tamper evident, blue polypropylene, linerless screw cap for narrow mouth/high form DIN thread soda lime glass bottles.
- Tamper evident ring
- LInerless
- High form DIN thread
Specifications
Specifications
| Description | Tamper-evident screw cap |
| For Use With (Application) | Storage of samples and reagents; Sample collection; Transport and shipping |
| Material | Polypropylene |
| Quantity | 10/Pk. |
| Thread Size (Metric) | GL 45 |
| Diameter (Metric) | 60 mm |
| Autoclavable | Autoclavable |