Learn More
Kautex™ Tamper-Evident Polypropylene Screw Closure
Click to view available options
Thread Size (Metric):
00 mm
Unit Size:
Pack of 550
Description
- Polypropylene construction
- Blue in color
- With tamper-evident ring and conical seal
Specifications
Specifications
| Color | Blue |
| For Use With (Application) | Providing Bottle Closure |
| Includes | Tamper evident ring, conical seal |
| Material | Polypropylene |
| Thread Size (Metric) | 00 mm |
| Neck Finish | 00 mm |
| For Use With (Equipment) | Series 310 Wide Neck Bottles 500 to 1000 mL Capacity |
| Closure Size | 54 cm |