Learn More
Brady™ Tamper-evident Metallized Thermal Transfer Printable Labels, 76 mm Core
Product Code. 11543764
Change view
Click to view available options
Height (Metric):
5.1 mm
6.4 mm
11.1 mm
25.4 mm
50.8 mm
Labels per Row:
1
4
5
Vertical Repeat (Metric):
7.6 mm
8.9 mm
13.6 mm
28.6 mm
54 mm
Width (Metric):
12.7 mm
16.5 mm
19.1 mm
50.8 mm
76.2 mm
Unit Size:
1 roll
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Height (Metric) Labels per Row Vertical Repeat (Metric) Width (Metric) unitSize
11543764 25.4 mm 1 28.6 mm 76.2 mm 1 roll
11593754 5.1 mm 4 7.6 mm 16.5 mm 1 roll
11503764 6.4 mm 4 8.9 mm 19.1 mm 1 roll
11533764 25.4 mm 1 28.6 mm 50.8 mm 1 roll
11513764 11.1 mm 5 13.6 mm 12.7 mm 1 roll
11553764 50.8 mm 1 54 mm 76.2 mm 1 roll
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 11543764 Supplier Brady™ Supplier No. 030183
Only null left
Add to basket
Add to basket
Printable, tamper-evident, metallized polyester labels for Thermal Transfer Printers.
  • Tamper-evident acrylic adhesive
  • For smooth surfaces
Specifications
Autoclavable Not Autoclavable
Certifications/Compliance UL Recognized
Adhesive Type Acrylic
Coating None
Color Silver
Finish Matte
Format Die-cut
Material Metallized Polyester (Tamper-evident)
Printable Sides Single-side Printable
Print Technology Thermal Transfer
Product Type Label
Sterility Non-sterile
For Use With (Application) Asset Tracking, Circuit Board Labeling, General Identificatio, Security Identification
For Use With (Equipment) BBP81, i5100, i7100, PR Plus, IP Series, THT 76 mm Core Printers, i7500
Labels per Row 1
Min. Operating Temperature (Metric) -40°C
Model THT-18-438-3
No. per Pack 3000 Label(s) / Roll
Quantity 3000 Labels
Shape Rectangle
Size 76.20 mm (W) x 25.40 mm (H)
Temperature Range (English) -40°F to +104°F
Temperature Range (Metric) -40°C to +40°C
Thickness (English) 0.0033 in.
Thickness (Metric) 0.084 mm
Vertical Repeat (Metric) 28.6 mm
Dimensions (L x W) 76.20 mm (W) x 25.40 mm (H)
Length (English) 1 in.
Width (English) 3 in.
Width (Metric) 76.2 mm
Width (English) Printable Area 76.20 mm
Width (Metric) Printable Area 76.2 mm
Height (English) 1 in.
Height (Metric) 25.4 mm
Height (Metric) Printable Area 25.4 mm
Alternate Part Number THT-18-438-3