Learn More
Köttermann Tall Safety Storage Cabinet with Drawers
Product Code. 17322139
Change view
Click to view available options
No. of Doors:
1
2
No. of Shelves:
4
5
8
9
10
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Doors No. of Shelves unitSize
17322139 1 4 Each
17332139 1 5 Each
17342139 2 8 Each
17362139 2 9 Each
17352139 2 10 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 17322139 Supplier Köttermann Supplier No. 37100036
This item requires a price update prior to being ordered.
Request a Price for this item here.
This Tall Safety Storage Cabinet offers top safety for flammable hazardous substances, complying with fire resistance class standards (Type 90).
  • Different versions that meet your needs
  • Additional storage space
  • Top safety for flammable hazardous substances (Type 90)
Specifications
Dimensions (D x W x H) 853 x 450 x 1966 mm
Door Style Drawer
Includes 1 Drip Tray
No. of Doors 1
No. of Shelves 4
Product Title
Selezionare un problema
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.