Learn More
Köttermann Tall Safety Storage Cabinet with Doors
Click to view available options
Door Style:
Folding, Manual-close
Hinged, Auto-close
No. of Shelves:
3
4
6
None
Includes:
1 Drip Tray
4 Internal Drawers
5 Internal Drawers
Unit Size:
Each
Description
- Different versions that meet your needs
- Additional storage space
- Top safety for flammable hazardous substances (Type 90)
Specifications
Specifications
| Dimensions (D x W x H) | 615 x 1200 x 1955 mm |
| Door Style | Hinged, Auto-close |
| Includes | 1 Drip Tray |
| No. of Doors | 2 |
| No. of Shelves | 6 |