Learn More
Agilent BioTek Take3 and Take3 Trio Micro-Volume Microplates
Take3 and Take3 Trio Micro-Volume plates expand functionality of the microplate spectrophotometers, multi-mode and Hybrid readers, while reducing sample volumes, costs and time required for routine quantification
3200.00€ - 5525.00€
Specifications
For Use With (Application) | Nucleic acid and protein quantification, in-situ fluorometric Pprotein quantification, 260/280 and 260/230 protein purity measurements, very low volume in-situ BCA assay, spectral scan of single or multiple low volume samples |
---|
Product Code | Brand | No. of Positions | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | No. of Positions | Price | Quantity & Availability | |||||
10626785
|
Agilent BioTek
TAKE3-SN |
16 |
3200.00€
Each |
Estimated Shipment: 24-06-2024
Log in to see stock. |
|||||
12568836
|
Agilent BioTek
TAKE3TRIO-SN |
48 |
5525.00€
Each |
Estimated Shipment: 24-06-2024
Log in to see stock. |
|||||
Description
Measure up to 16 (Take3) or 48 (Take3 Trio) 2μL samples at a time without dilution or special treatment.
- Compatible with Agilent BioTek Eon, Epoch, Synergy, PowerWave XS2 spectrophotometers
- Includes Gen5 software with pre-programmed micro-volume nucleic acid and protein quantification protocols
- Absorbance and fluorescence detection capability
- Easy maintenance: simply wipe-off sample locations
- Robust design
- Take3 Trio has two locations for BioCell measurements
- Read microspots, BioCells or standard cuvets
Take3 (11-120-571) offers:
- 16 microspots for 2μL samples; two locations for proprietary BioCell measurements (BioCell optional) and one location for standard cuvet measurements
- 48 microspots for 2μL samples; two locations for proprietary BioCell measurements (BioCell optional)
Nucleic acid and protein quantification, in-situ fluorometric protein quantification, 260/280 and 260/230 protein purity measurements, very low volume in-situ BCA assay, spectral scan of single or multiple low volume samples
Specifications
Nucleic acid and protein quantification, in-situ fluorometric Pprotein quantification, 260/280 and 260/230 protein purity measurements, very low volume in-situ BCA assay, spectral scan of single or multiple low volume samples |