Learn More
TAK-875, TRC
Product Code. 17903685
Change view
Click to view available options
Quantity:
5 mg
25 mg
50 mg
Unit Size:
25mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17903685 25 mg 25mg
17953725 5 mg 5mg
17943945 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 17903685 Supplier Toronto Research Chemicals Supplier No. TRCT00491525MG
 Restricted Item
Estimated Shipment: 14-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1000413-72-8
Molecular Formula C29 H32 O7 S
Molecular Weight (g/mol) 524.6252
Synonym (3S)-6-[[2',6'-Dimethyl-4'-[3-(methylsulfonyl)propoxy][1,1'-biphenyl]-3-yl]methoxy]-2,3-dihydro-3-benzofuranacetic Acid, (+)-TAK 875, TAK-875
IUPAC Name 2-[(3S)-6-[[3-[2,6-dimethyl-4-(3-methylsulfonylpropoxy)phenyl]phenyl]methoxy]-2,3-dihydro-1-benzofuran-3-yl]acetic acid
SMILES Cc1cc(OCCCS(=O)(=O)C)cc(C)c1c2cccc(COc3ccc4[C@H](CC(=O)O)COc4c3)c2
Specifications
Quantity 25 mg
InChI Formula InChI=1S/C29H32O7S/c1-19-12-25(34-10-5-11-37(3,32)33)13-20(2)29(19)22-7-4-6-21(14-22)17-35-24-8-9-26-23(15-28(30)31)18-36-27(26)16-24/h4,6-9,12-14,16,23H,5,10-11,15,17-18H2,1-3H3,(H,30,31)/t23-/m1/s1
Formula Weight 524.1869
Chemical Name or Material 2-[(3S)-6-[(3-[2,6-dimethyl-4-(3-methylsulfonylpropoxy)phenyl]phenyl)methoxy]-2,3-dihydro-1-benzofuran-3-yl]acetic acid