missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ TAK 715
Description
Inhibitor of p38 MAPK (IC50 = 7.1 nM for p38 MAPKα). Also inhibits Wnt-3a-stimulated β-catenin signaling. Inhibits 22 kinases by more than 80%, including CK1δ/ε. Anti-inflammatory and anti-rheumatoid arthritis agent. Orally bioavailable.
Specifications
Specifications
| Quantity | 50 mg |
| Product Line | Tocris |
| Formula Weight | 399.51 |
| Purity | >99% |
| Molecular Weight (g/mol) | 399.512 |
| Chemical Name or Material | TAK 715 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?