missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ TAK 375
GREENER_CHOICE

Product Code. 18728586
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18728586

Brand: Tocris Bioscience™ 6777/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Very high affinity and selective melatonin receptor agonist

TAK 375 is a very high affinity and selective melatonin agonist (Ki values are 14 and 112 pM for human MT1 and MT2, respectively). Selective for human MT1 and MT2 over hamster MT3 and a range of other targets including benzodiazepine, opiate and dopamine receptors, ion channels and transporters. Accelerates return to normal circadian rhythm in rats after phase shift, without affecting learning and memory. Also reduces infarct size in rat heart in vitro post ischemia-reperfusion.
TRUSTED_SUSTAINABILITY

Specifications

Synonym Ramelteon
Quantity 10 mg
Molecular Formula C16H21NO2
Formula Weight 259.34
Product Type Agonist
Purity 98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.