missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TAFA3/FAM19A3 Mouse anti-Human, Alexa Fluor 405, Clone: 460904, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems IC4677V-100UG
This item is not returnable.
View return policy
Description
TAFA3/FAM19A3 Monoclonal specifically detects TAFA3/FAM19A3 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
| TAFA3/FAM19A3 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
| Q7Z5A8 | |
| FAM19A3 | |
| E. coli-derived recombinant human TAFA3/FAM19A3 Ala31-Arg133 Accession # Q7Z5A8 | |
| RUO | |
| 284467 | |
| Human | |
| IgG2b |
| Flow Cytometry | |
| 460904 | |
| Alexa Fluor 405 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| Chemokine-like protein TAFA-3, FAM19A3, family with sequence similarity 19 (chemokine (C-C motif)-like), member A3, MGC138473, TAFA3, TAFA-3 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human TAFA3/FAM19A3 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human TAFA1, 2, 4, or 5 is observed. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
For Research Use Only (RUO)