Learn More
Testo™ Tachometer
Product Code. 10106211
Change view
Click to view available options
Range:
1 to 99,999 rpm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Range Unit Size
10106211 1 to 99,999 rpm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10106211 Supplier Testo™ Supplier No. 0563 0465
  • ONLINE EXCLUSIVE PRICE
    509.12€
    243.00€ / Pack of 1
    Save 266.12€
    52% Off
 Restricted Item
Estimated Shipment: 07-12-2026
Specifications
Product Type Tachometer
Includes Plastic transport case, SoftCase protective case, reflective markers, and batteries
Range 1 to 99,999 rpm