Learn More
(-)-Tabersonine Hydrochloride, TRC
Specifications
Specifications
| Chemical Name or Material | (-)-Tabersonine Hydrochloride |
| CAS | 29479-00-3 |
| Molecular Formula | C21H24N2O2 HCl |
| Quantity | 5 mg |
| Synonym | (5α,12R,19α)-2,3,6,7-Tetradehydroaspidospermidine-3-carboxylic Acid Methyl Ester Hydrochloride, 3a-Ethyl-3a,4,6,11,12,13a-hexahydro-1H-indolizino[8,1-cd]carbazole-5-carboxylic Acid Methyl Ester Hydrochloride, Tabersonine Hydrochloride, Tabersonin Hydrochloride |
| InChI Formula | InChI=1S/C21H24N2O2.ClH/c1-3-20-9-6-11-23-12-10-21(19(20)23)15-7-4-5-8-16(15)22-17(21)14(13-20)18(24)25-2;/h4-9,19,22H,3,10-13H2,1-2H3;1H/t19-,20-,21-;/m0./s1 |
| SMILES | Cl.CC[C@@]12CC(=C3Nc4ccccc4[C@@]35CCN(CC=C1)[C@@H]25)C(=O)OC |
| Molecular Weight (g/mol) | 372.9 |
| Formula Weight | 372.1605 |