Learn More
(-)-Tabersonine Hydrochloride, TRC
Product Code. 30410820
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30410820 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30410820 Supplier Toronto Research Chemicals Supplier No. TRCT0039051MG
 Restricted Item
Estimated Shipment: 12-11-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material (-)-Tabersonine Hydrochloride
CAS 29479-00-3
Molecular Formula C21H24N2O2 HCl
Quantity 1 mg
Synonym (5α,12R,19α)-2,3,6,7-Tetradehydroaspidospermidine-3-carboxylic Acid Methyl Ester Hydrochloride, 3a-Ethyl-3a,4,6,11,12,13a-hexahydro-1H-indolizino[8,1-cd]carbazole-5-carboxylic Acid Methyl Ester Hydrochloride, Tabersonine Hydrochloride, Tabersonin Hydrochloride
InChI Formula InChI=1S/C21H24N2O2.ClH/c1-3-20-9-6-11-23-12-10-21(19(20)23)15-7-4-5-8-16(15)22-17(21)14(13-20)18(24)25-2;/h4-9,19,22H,3,10-13H2,1-2H3;1H/t19-,20-,21-;/m0./s1
SMILES Cl.CC[C@@]12CC(=C3Nc4ccccc4[C@@]35CCN(CC=C1)[C@@H]25)C(=O)OC
Molecular Weight (g/mol) 372.9
Formula Weight 372.1605
Título del producto
Seleccione un problema
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.