Learn More
Thermo Scientific™ T-valve Assembly for LN2 Supply Tank
Product Code. 15152889 Shop All Thermo Scientific Products
Click to view available options
:
Manual valve and T fitting
Product Code. 15152889
Brand: Thermo Scientific™ 4000290
Connect two hoses to a LN2 tank with this T-valve assembly, which includes a manual valve and T-fitting.
Connect two hoses to a LN2 tank with this Thermo Scientific™ T-valve assembly for LN2 supply tank, which includes a manual valve and T-fitting.
TRUSTED_SUSTAINABILITY
Specifications
Description Manual valve and T fitting
Type Manual Valve and T-Fitting
For Use With (Equipment) CryoPlus 1,2,3,4
Defrost Manual