missing translation for 'onlineSavingsMsg'
Learn More

T 26c disodium salt

Product Code. 18788396
Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
This item is not returnable. View return policy

Product Code. 18788396

Brand: Tocris Bioscience 6134/5

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products

This item is not returnable. View return policy

Highly potent and selective MMP13 inhibitor

T 26c disodium salt is a highly potent and selective MMP13 inhibitor (IC50 = 6.9 pM). Exhibits >2600-fold selectivity for MMP13 over related MMPs. Inhibits degradation of bovine nasal septum cartilage explants in vitro . Orally bioavailable.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Disodium 4-[[[[2-[[[[3-(Methyloxy)phenyl]methyl]amino]carbonyl]-4-oxo-3,4-dihydrothieno[2,3-d]pyrimidin-5-yl]methyl]oxy]methyl]benzoate
CAS 869298-22-6
Assay Percent Range >98%
Quantity 5 mg
Molecular Formula C24H19N3Na2O6S
Formula Weight Observed MW: 523.47
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.