Learn More
CELLPATH SYSTEM III MICRO BIOPSY CASSETTE
Product Code. p-200113601
Click to view available options
Material:
Loose
Threaded
Unit Size:
Pack of 2000
Pack of 500
Product Code. 30226238
Brand: CELLPATH EAI070210A
The System III Micro Biopsy Cassette comes with an attached lid with an easy to break-off hinge. Incorporating square apertures of 0.38 mm by 0.38 mm, the Micro Biopsy Cassette ensures safe accommodation of smaller biopsy specimens, whilst also enabling good levels of fluid transfer.
  • The locking mechanism of the cassette ensures maximum tissue security during processing.
  • The cassette has a precisely engineered writing surface which optimises the print quality on inkjet and thermal printers, including the Primera, Leica and Sakura Printers.
  • Key Benefits:
  • Compatible with Primera, Leica and Sakura Printers
  • Precisely engineered writing surface
  • Increased reagent flow
  • Integral fixed frangible lid
  • High rigidity and stability
Specifications
Color White
Material Loose
Product Type Cassette