Learn More
CELLPATH SYSTEM II+ MICRO BIOPSY CASSETTE - WHITE LASER (THREADED)
Product Code. 30226254
Change view
Click to view available options
Color:
White
Unit Size:
Pack of 2000
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
30226254 White Pack of 2000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30226254 Supplier CELLPATH Supplier No. EAM150272B
  • ONLINE EXCLUSIVE PRICE
    407.61€
    245.96€ / Pack of 2000
    Save 161.65€
    39% Off
 Restricted Item
Estimated Shipment: 04-12-2026
The System II+ Micro Biopsy Laser Cassette incorporates an inner chamber with 0.38 x 0.38 mm apertures. The inner chamber has rounded corners that enable easy handling of small samples using forceps.
  • Read rates
  • Robust and secure closure ensures specimen security
  • Designed to improve reagent flow
  • Large color range
  • Writing surface suitable for marking with a CellMark pen
Specifications
Color White
Product Type Cassette