Learn More
Thermo Scientific™ Syringes for Thermo Scientific™ HPLC Instruments
Product Code. 10782127 Shop All Thermo Scientific Products
Change view
Click to view available options
Volume (Metric):
250 μL
500 μL
2.5 mL
For Use With (Equipment):
AS1000, AS3000 Autosampler
AS3000, AS3500 Autosampler
LCQ HPLC
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) For Use With (Equipment) Unit Size
10782127 250 μL LCQ HPLC Each
10341205 500 μL LCQ HPLC Each
10702917 250 μL AS1000, AS3000 Autosampler Each
10321065 500 μL AS1000, AS3000 Autosampler Each
10197744 2.5 mL AS3000, AS3500 Autosampler Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 10782127 Supplier Thermo Scientific™ Supplier No. 365ILT21
Save Up to 60% on Contract Testing Essentials
Use promo code 28025
View all promotions
Estimated Shipment: 21-09-2026
Add to basket
Add to basket
Thermo Scientific™ Syringes for Thermo Scientific HPLC Instruments offer easy, accurate and reproducible injections.
Thermo Scientific™ Syringes for Thermo Scientific HPLC Instruments offer easy, accurate and reproducible injections.
  • Square tip to prevent damage to the injector
  • Wide range of volumes
  • Precision made from borosilicate glass and stainless steel
  • Robust design and easy-to-read markings
TRUSTED_SUSTAINABILITY
Specifications
Needle Gauge 22 G
Length (English) Needle 2 in. Removable Needle
Product Type HPLC Autosampler Syringe
Disposable Reusable
Volume (Metric) 250 μL
For Use With (Equipment) LCQ HPLC