Learn More
Hamilton™ Custom Cemented Needle Syringes
The gas-tight version of the original 700 series syringes.
85.00€ - 1615.00€
Product Code | Brand | Volume (Metric) | Product Type | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Product Type | Price | Quantity & Availability | ||||
11518024
|
Hamilton™
DS80008 |
10 μL | Gastight |
1615.00€
Each |
Estimated Shipment: 06-05-2024
Log in to see stock. |
||||
10595705
|
Hamilton™
201550 |
100 μL | Syringe Needle |
201.00€
Each |
Estimated Shipment: 03-06-2024
Log in to see stock. |
||||
10194402
|
Hamilton™
81008 |
100 μL | Gastight |
171.00€
Each |
Estimated Shipment: 06-05-2024
Log in to see stock. |
||||
10491245
|
Hamilton™
80208 |
25 μL | Gastight |
145.00€
Each |
Estimated Shipment: 06-05-2024
Log in to see stock. |
||||
10281973
|
Hamilton™
202134 |
250 μL | Syringe Needle |
91.00€
Each |
Estimated Shipment: 03-06-2024
Log in to see stock. |
||||
10201242
|
Hamilton™
81108 |
250 μL | Gastight |
171.00€
Each |
Estimated Shipment: 06-05-2024
Log in to see stock. |
||||
10234272
|
Hamilton™
82525 |
25 mL | Syringe Needle |
251.00€
Each |
Estimated Shipment: 03-06-2024
Log in to see stock. |
||||
10175122
|
Hamilton™
80908 |
50 μL | Gastight |
179.00€
Each |
Estimated Shipment: 06-05-2024
Log in to see stock. |
||||
10349551
|
Hamilton™
202193 |
500 μL | Syringe Needle |
85.00€
Each |
Estimated Shipment: 27-05-2024
Log in to see stock. |
||||
Description
- Ideal for dispensing volumes from 1 up to 500 μL
- A variety of volumes, lengths, and gauges available
Specifications
Not Autoclavable | |
1 Ea. |