Learn More
Syngene™ Visible Light Converter
Product Code. 12804068 Shop All Syngene Products
Change view
Click to view available options
For Use With (Equipment):
G:BOX Systems
IG/UG and DG Systems
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
12804068 G:BOX Systems Each
12894058 IG/UG and DG Systems Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 12804068 Supplier Syngene™ Supplier No. GXCONVERT5
Estimated Shipment: 21-12-2026
Use the Visible Light Converter in conjunction with your UV transilluminator. Screen provides a white surface for visualising visible protein gels.
  • Use the Visible light converter in conjunction with a UV transilluminator provides a white surface for visualising visible protein gels.
Specifications
Intensity Fixed
Wavelength 302 nm
Description UV (White)
For Use With (Equipment) G:BOX Systems
Type Converter Plate