Learn More
BOLA Swivelling Screw Fittings with Ground Joint
Click to view available options
Diameter (Metric):
6 mm
10 mm
Unit Size:
Each
Description
- Ground joint body made of PTFE, screw cap made of PPS.
- Probes, tubes or tubing are firstly inserted through the fitting, then the fitting is put into the ground joint and the inserted element can be fixed in the requested position.
- Suitable for all kinds of probes, tubes or hard-walled tubing
- Can be used with GL threads or ground joints
- Angle and height can be fixed tightly
- A damage of tube or probe due to collision with the stirrer shaft can be avoided
Specifications
Specifications
| Certifications/Compliance | FDA |
| Diameter (Metric) | 6 mm |
| Autoclavable | Autoclavable |
| Arm Angle | 8° |
| Color | White |
| Flexibility | Rigid |