Learn More
Notrax Swisslon XT™ Premium Quality Barrier Matting, Mocha
Product Code. 17792038
Click to view available options
Length (Metric):
90 cm
120 cm
150 cm
180 cm
240 cm
Unit Size:
Each
Product Code. 17792038
Brand: Notrax 380S0048BR
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Notrax Swisslon XT™ Premium quality barrier matting designed to remove walked-in moisture and finer dirt particles in large indoor entrance areas, including wall-to-wall applications.
  • Premium quality barrier matting designed to remove walked-in moisture and finer dirt particles in large indoor entrance areas, including wall-to-wall applications
  • Our redesigned classic barrier mat, known for its softer yarns, has been enhanced with a mono-filament to offer additional scraping and provide excellent cleaning and absorbing
  • The new tri-color grit pattern provides outstanding dirt-hiding characteristics
  • Great ability to retain moisture (<58 liters per m²) and scrape debris from foot traffic while maintaining its great looking appearance for the longest time
  • A dense heavyweight vinyl backing ensures longevity
Specifications
Dimensions (L x W) 240 x 120 cm
Color Mocha
Length (English) 94 in.
Length (Metric) 240 cm
Material Polyamide 6.0 Yarns
Type Industrial and Commercial Matting
Width (Metric) 120 cm