Learn More
Eppendorf™ Swing Out Rotor for Benchtop Centrifuge
Features a compact and robust design, and superior operating comfort
Brand: Eppendorf™ 5702740004
611.00 EUR valid until 2024-12-31
Use promo code "23292" to get your promotional price.
Additional Details : Weight : 1.30000kg
Description
- For use with Eppendorf Centrifuges 5702
- 5702 family of low speed, general purpose centrifuges was especially designed to meet the requirement of low- to mid-throughput clinical research and cell culture labs. They are compact in design so that one can fit comfortably on every lab bench, quiet in operation to benefit your work environment.
Specifications
Swing-out Rotor without Basket | |
3,000 x g | |
5702/5702 R/5702 RH Centrifuge |
5 to 25 mL | |
4,400 rpm | |
15 |