Learn More
Hettich™ Swing-out Rotor for ROTOFIX 32 A and Universal 320/320 R Centrifuges, 4-place
Click to view available options
Max. Speed:
4,000 rpm
Unit Size:
Each
Description
- Swing-out rotor, 4 place
- With biocontainment
- For use with Hettich Centrifuges
- Universal 320/320 R Centrifuges feature excellent performance and a comprehensive range of accessories
- ROTOFIX Centrifuges are ideally equipped for tasks in clinical chemistry, it can also play a vital role in cytology
Specifications
Specifications
| Type | Swing-out Rotor |
| Max. RCF | 3,328 x g |
| Max. Speed | 4,000 rpm |
| Radius (Metric) | 147 mm |
| Radius (Metric) Min. | 133 mm |
| Max. Tube Length | 100 mm |
| Max. Tube Diameter | 44 mm |
| For Use With (Equipment) | ROTOFIX 32 A and Universal 320/320 R Centrifuges |
| Angle | 90° |
| No. of Places | 4 |
| Show More |