Learn More
Hanna Instruments™ Swimming Pool Chemical Test Kit
Click to view available options
Range:
0 to 2.5 mg/L
Unit Size:
Pack of 150
Specifications
Specifications
| Quantity | 1 Each |
| Range | 0 to 2.5 mg/L |
| Test Method | DPD |
| Type | Swimming Pool Chemical Test Kit |
Safety and Handling
Product Identifier
- X150 TEST KIT CHLORINE LIBRE PH
- Danger
- Serious eye damage/eye irritation Category 1
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 A
- Specific target organ toxicity after repeated exposure Category 2
- H290-May be corrosive to metals.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H373-May cause damage to organs.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P390-Absorb spillage to prevent material damage.
- MIXTURE LIST-Contains : sodium hydroxide, sulphuric acid, ethanediol