Learn More
SVL™ Torion Right Angled Stopcocks
Product Code. 15653447
Click to view available options
Plug Bore:
14.7 mm, Side Arm 10 mm
5.7 mm, Side Arm: 3 mm
8.7 mm, Side Arm: 5 mm
Unit Size:
Pack of 10
Pack of 5
Product Code. 15653447
Brand: SVL™ 72242
Only null left
Add to basket
Add to basket
Manufactured from borosilicate glass
  • PTFE key is screwed into the threaded Torion connector, to open and close the stopcock
  • No lubrication required, no seizure and no contamination
  • Vacuum capability to 10-2 Torr
  • Sidearms are 70mm in length
TRUSTED_SUSTAINABILITY
Specifications
Type Right Angle
Plug Bore 8.7 mm, Side Arm: 5 mm
Material Borosilicate Glass
Diameter (Metric) Side Arm 8 mm