Learn More
SVL™ Sealing Ring for Butt Joint
Description
- Silicone rubber and sheathed with PTFE for protection against chemical attack
- Internal diameter of ring corresponds to the bore of the SVL glass screwthread so that flow is not restricted
Specifications
Specifications
| Description | Joint size: 30 |
| For Use With (Application) | Butt joints |
| Joint Size | 30 |
| Material | Silicone Rubber, PTFE (Sheath) |
| Type | Seal Ring |