Learn More
Quickfit™ Claisen Still Head Adapter
Product Code. p-8003027
Click to view available options
Joint Type:
Condenser: 19/26, Flask: 24/29
Condenser: 24/29, Flask: 24/29
Size:
Socket: 14/23
Unit Size:
Each
Pack of 5
Product Code. 12048493
Brand: Quickfit SH6/33
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
PTFE joint sleeves eliminate the possibility of joint seizure
  • Similar to the SH4: still heads but with extra socket to accept dropping funnel or stirrer
TRUSTED_SUSTAINABILITY
Specifications
Type Claisen Still Head
Material Borosilicate Glass
Standard Taper Joint 24/49 (Condenser), 24/29 (Flask)
Joint Type Condenser: 24/29, Flask: 24/29
Size Socket: 14/23