Learn More
Novus Biologicals™ Surfactant Protein A Overexpression Lysate
Product Code. 18096428 Shop All Bio Techne Products
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.10mg
Product Code. 18096428
Brand: Novus Biologicals™ NBP211484
The Surfactant Protein A Overexpression Lysate is derived from human. It has been validated for the following applications: Western Blot.
TRUSTED_SUSTAINABILITY
Specifications
Cell Line The lysate was created in HEK293T cells, using plasmid ID RC215316 and based on accession number NP_1158116. The protein contains a C-terminal DDK tag.
For Use With (Application) Western Blot
Quantity 0.1 mg
Storage Buffer RIPA buffer
Purity Protein
Test Specificity Homo sapiens Surfactant Protein A, mRNA.
Content And Storage -80°C. Avoid freeze-thaw cycles.
Protein State Native