Learn More
Novus Biologicals™ Surfactant Protein A Antibody Pair
Product Code. 18537290 Shop All Bio Techne Products
Click to view available options
Quantity:
1 Set
Unit Size:
1 set
Product Code. 18537290
Brand: Novus Biologicals™ H00653509AP43
Specifications
Assay Range 3 ng/mL to 100 ng/mL
Conjugate Unconjugated
Product Type Sandwich ELISA
For Use With (Application) Sandwich ELISA
Gene ID (Entrez) 653509
Gene Symbol SFTPA1
Quantity 1 Set
Storage Requirements Aliquot and store at -20°C to +80°C. Avoid freeze/thaw cycles.
Synonym COLEC4, PSAP, PSPA, PSP-A, SFTP1, SFTPA1B, SPA, SP-A, SPA1, SP-A1, surfactant protein A1B
Target Human
Buffer 1 x PBS, pH 7.4