Learn More
Dostmann Electronic™ Surface Probe
Product Code. 11854302
Click to view available options
Diameter (Metric) Probe:
26 mm
Unit Size:
Each
Product Code. 11854302
Brand: Dostmann Electronic™ 69100070
Specifications
Diameter (Metric) Probe 26 mm
For Use With (Application) For Measurement of Inaccessible Surfaces and/or Moving Surface.
Product Type Surface Probe
For Use With (Equipment) HiTemp 1800 Infrared thermometer, HiTemp 2400 Infrared thermometer.Mini-K industry thermometer, P410 Profi Thermometer Thermocouple Type-K Proscan 520 Infrared thermometer, Proscan 530 Infrared thermometer, RH 896 Infrared thermometer with hygrometer., Sc