Description
- Kills 99.99% of enveloped viruses in five minutes including Coronavirus EN14476: Limited virucidal activity (active against enveloped viruses such as, e.g. Influenza Virus, Coronavirus, HIV, Herpes Virus, Hepatitis Virus) exposure time: 5 minutes under dirty conditions.
- Biodegradable formula. 100% plant-based active ingredient (lactic acid).
- Food surface contact safe The Surface Disinfecting Spray is suitable for PT2 (disinfectants not intended for direct application to humans or animals) and PT4 (food and feed area) applications, food contact surfaces must be rinsed with drinking water after disinfecting with product.
- The product is an alcohol-free ready-to-use multi-purpose spray for surface disinfection.
Specifications
Specifications
| Product Type | Trigger Spray Bottle |
| Color-coded | No |
| Form | Clear Liquid |
| Scent | Unscented |
| Sterility | Non-sterile |
| Color | Clear |
| Packaging | Plastic Bottle |
| Alternate Part Number | 7322541432632 |