Description
- Telescopic samplers are often used for taking surface samples from rivers, tanks and lakes
- A wide choice of attachments is available to make the TeleScoop ideally suited for most applications
- Attachments are interchangeable and simply click into position
Specifications
Specifications
| Product Type | Swabbing Template |
| Material | Polypropylene |
| Autoclavable | Not Autoclavable |
| Disposable | Reusable |
| Graduated | No |
| Sterility | Non-sterile |