Learn More
Thermo Scientific™ Support Stands for Thermo Scientific™ Heratherm Ovens and Incubators
Product Code. 10304130 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Equipment):
100 L Model Ovens and Incubators
180 L Model Ovens and Incubators
60 L Model Ovens and Incubators
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
10304130 100 L Model Ovens and Incubators Each
10811311 60 L Model Ovens and Incubators Each
10724602 180 L Model Ovens and Incubators Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10304130 Supplier Thermo Scientific™ Supplier No. 50127742
Support stands with casters
Height including casters: 7.36 in. (18.7cm).
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) 100 L Model Ovens and Incubators
Type Support Stand
Includes Caster