Learn More
Thermo Scientific SUPPORT FRAME 780mm HIGH 50045014
Brand:  Thermo Scientific 50045014
SUPPORT FRAME 780mm HIGH 50045014
Product Code. 10006551
  • ONLINE EXCLUSIVE PRICE
    1045.00€
    989.00€ / Pack of 1
    Save 56.00€
    5% Off
 Restricted Item
Estimated Shipment: 25-01-2027