Learn More
Epredia™ SuperFrost™ Microscope Slides, Ground 90°
Product Code. 12392098 Shop All Epredia Products
Change view
Click to view available options
Color:
Blue
Green
Pink
White
Yellow
Quantity:
50/Box, 2000 in outer carton
Unit Size:
Pack of 50
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Quantity unitSize
12392098 Pink 50/Box, 2000 in outer carton Pack of 50
12372098 White 50/Box, 2000 in outer carton Pack of 50
12302108 Yellow 50/Box, 2000 in outer carton Pack of 50
12382098 Blue 50/Box, 2000 in outer carton Pack of 50
12312108 Green 50/Box, 2000 in outer carton Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 12392098 Supplier Epredia™ Supplier No. AG00008232E01FST20
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
The printed Epredia™ SuperFrost tab is impervious to virtually all common laboratory chemicals and reagents. A wide range of colors (7 standard colors) allows several examination specimens to be labeled and archived simply and clearly.
  • Time-consuming labeling with paper labels is no longer required
  • The writing area is resistant to all common solvents
  • SuperFrost slides may be sorted by color e.g. by patient, urgency, stain or examination method
  • Simple recognition of specified characteristics such as year of examination or department, use etc.
TRUSTED_SUSTAINABILITY
Tekniske data
Length (Metric) 76 mm
Width (Metric) 26 mm
Edges 90°, Ground
Thickness (Metric) 1 mm
Color Pink
Certifications/Compliance ISO8037/I
Coating Uncoated
Quantity 50/Box, 2000 in outer carton
Writing Surface Yes
Dimensions (L x W) 76 x 26 mm
Produkttitel
Vælg et problem
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.