Learn More
Epredia™ SuperFrost™ Plus Slides
Product Code. 12608106 Shop All Epredia Products
Change view
Click to view available options
:
White; 144/cs
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
12608106 White; 144/cs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12608106 Supplier Epredia™ Supplier No. 6776214CS
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Epredia™ Superfrost™ Plus Slides are positively charged for improved cellular adhesion.
  • Electrostatically attract tissue sections and cytology preparations, binding them to the slide
  • Eliminate the need for special adhesives or protein coatings
  • No background staining
  • Available in a variety of colors
TRUSTED_SUSTAINABILITY
Specifications
Frosted Yes
Type Microscope Slide
Color White
Quantity 1440/Cs.