Få mere at vide
Metro™ Super Adjustable Super Erecta™ Wire Shelf, Chrome Finish
Produktkode 11765669
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Width (English):
14 in.
18 in.
21 in.
24 in.
Length (English):
24 in.
30 in.
36 in.
42 in.
48 in.
54 in.
60 in.
72 in.
Förpackningsstorlek
Each
20 product options available for selection
Product selection table with 20 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkode Width (English) Length (English) unitSize
11765669 24 in. 60 in. Each
15946851 24 in. 54 in. Each
15946821 18 in. 60 in. Each
15946841 24 in. 30 in. Each
15916811 18 in. 30 in. Each
15966811 18 in. 42 in. Each
15916821 18 in. 54 in. Each
15906841 21 in. 72 in. Each
15946831 21 in. 36 in. Each
15966831 21 in. 48 in. Each
11745669 24 in. 36 in. Each
15996841 24 in. 42 in. Each
11755669 24 in. 48 in. Each
15976821 18 in. 72 in. Each
15986831 21 in. 60 in. Each
15906831 21 in. 24 in. Each
15906801 14 in. 36 in. Each
11735669 18 in. 48 in. Each
11725669 18 in. 36 in. Each
15926841 24 in. 24 in. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 20 options available.
20 missing translation for 'options'
Denne vare kan ikke returneres. Se returpolitik
Produktkode 11765669 missing translation for 'mfr' Metro™ missing translation for 'supplierNo' A2460NC
 Restricted Item
Estimated Shipment: 27-11-2026
Tilføj til indkøbskurv
Tilføj til indkøbskurv
Denne vare kan ikke returneres. Se returpolitik
Indeholder
Each shelf shipped with one pack of four nonconductive sleeve/wedges.
Unique, open-wire design of chrome finish shelf permits many configurations, free circulation of air, and minimizes dust accumulation.
  • Each shelf provides approximately 76% open area for increased light penetration and improved visibility of stored items.
  • Can be loaded and unloaded easily from all sides.
  • Hard, durable surface.
  • Abrasion resistant.
  • Ideal for dry, noncorrosive environments.
  • Shelves adust in 1 in. increments with patented shelf-release corners.
  • Shelf releases and wedges offer improved rigidity.
  • Assembles quickly without tools; assembled units are nonconductive.
Ordreinfo
Posts sold separately.
Tekniske data
Product Type Wire Shelf
Dimensions (L x W) 60 x 24 in. (152.4 x 61 cm)
For Use With (Application) Ideal for Drying, Non-corrosive Environments
For Use With (Equipment) Metro™ Super Erecta™ adjustable shelving system
Includes Four non conductive sleeve/wedges
Length (English) 60 in.
Length (Metric) 152.4 cm
Width (English) 24 in.
Width (Metric) 61 cm
Material Chrome
Produkttitel
Vælg et problem
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.