Learn More
Metro™ Super Adjustable Super Erecta™ Wire Shelf, Chrome Finish
Product Code. 11725669
Click to view available options
Width (English):
14 in.
18 in.
21 in.
24 in.
Length (English):
24 in.
30 in.
36 in.
42 in.
48 in.
54 in.
60 in.
72 in.
Unit Size:
Each
Product Code. 11725669
Brand: Metro™ A1836NC
Only null left
Add to basket
Add to basket
Includes
Each shelf shipped with one pack of four nonconductive sleeve/wedges.
Unique, open-wire design of chrome finish shelf permits many configurations, free circulation of air, and minimizes dust accumulation.
  • Each shelf provides approximately 76% open area for increased light penetration and improved visibility of stored items.
  • Can be loaded and unloaded easily from all sides.
  • Hard, durable surface.
  • Abrasion resistant.
  • Ideal for dry, noncorrosive environments.
  • Shelves adust in 1 in. increments with patented shelf-release corners.
  • Shelf releases and wedges offer improved rigidity.
  • Assembles quickly without tools; assembled units are nonconductive.
Order Info
Posts sold separately.
Specifications
Product Type Wire Shelf
Dimensions (L x W) 36 x 18 in. (91.4 x 45.7 cm)
For Use With (Application) Ideal for Drying, Non-corrosive Environments
For Use With (Equipment) Metro™ Super Erecta™ adjustable shelving system
Includes Four non conductive sleeve/wedges
Length (English) 36 in.
Length (Metric) 91.4 cm
Width (English) 18 in.
Width (Metric) 45.7 cm
Material Chrome