Learn More
Diversey™ Suma Alcohol Wipes
Product Code. 17603095
Change view
Click to view available options
Includes:
Wipes for AT, DE, FR, GB, IE only
Unit Size:
Pack of 1800
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Includes Unit Size
17603095 Wipes for AT, DE, FR, GB, IE only Pack of 1800
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17603095 Supplier Diversey™ Supplier No. 100878518
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
No-rinse alcoholic disinfectant wipes
Suma Alcohol Wipes are ready-to-use wipes for disinfection of all hard surfaces like work areas, small equipment, utensils, and handholds.
Key properties
  • Suma Alcohol Wipes are perfect for disinfection of all food contact areas and can be used as complement to your cleaning and disinfection procedures.
  • The product is based on ethanol to provide fast and effective disinfection.
  • It is suitable for the disinfection of alcohol-safe, food contact surfaces and articles such as chopping boards, utensils, thermometers, tables and other smaller kitchen equipment.
  • It is effective against a wide spectrum of micro-organisms.
Benefits
  • Easy to use
  • No spraying, safe to use
  • Effective against a wide range of micro-organisms
  • Quick drying minimizes disruption to work flow
Specifications
Product Type Alcohol Wipes
Length (Metric) 135 mm
Width (Metric) 180 mm
Dimensions (L x W) 135 x 180 mm
Packaging Quantity 12 x 150 Wipes
Includes Wipes for AT, DE, FR, GB, IE only
compliance-icons compliance-icons
Product Identifier
  • X1800 Suma Alcohol Wipe, 150/pack, 12 packs - AT, DE, FR, GB & IE only
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P403+P235-Store in a well-ventilated place. Keep cool.
Supplemental information
  • MIXTURE LIST-Contain: ethanol, propan-2-ol